Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: MEFV | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000243.2 | NP_000234.1 | 3 | 1006 | ||
NM_001198536.1 | NP_001185465.1 | 2 | 373 | |||
XM_017023236.1 | XP_016878725.1 | 3 | 1147 | |||
GenBank mRNA | AF018080.1 | - | 3 | 1007 | ||
AK298660.1 | - | 3 | 1000 | |||
BC101511.1 | - | 3 | 991 | 8069017 | ||
BC101537.1 | - | 3 | 991 | 8069043 | ||
FJ785722.1 | - | 2 | 333 | |||
Y14441.1 | - | 3 | 995 |