Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PEX7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000288.3 | NP_000279.1 | 4 | 480 | ||
XM_005267019.4 | XP_005267076.1 | 4 | 760 | |||
XM_017010934.1 | XP_016866423.1 | 4 | 824 | |||
GenBank mRNA | BC006268.1 | - | 4 | 422 | 3948721 | |
BC031606.1 | - | 4 | 464 | 5176358 | ||
CR542054.1 | - | 4 | 378 | |||
CR542076.1 | - | 4 | 378 | |||
U76560.1 | - | 4 | 426 | |||
U88871.1 | - | 4 | 459 |