Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDH5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK300461.1 | - | 3 | 478 | ||
BC096364.3 | - | 3 | 444 | 40012045 | ||
BC096365.1 | - | 3 | 444 | 40012047 | ||
DC381809.1 | - | 3 | 540 | |||
U84722.1 | - | 3 | 502 | |||
X79981.1 | - | 2 | 407 |