Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TYR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000372.4 | NP_000363.1 | 2 | 972 | ||
XM_011542970.2 | XP_011541272.1 | 2 | 1279 | |||
GenBank mRNA | AB775899.1 | - | 2 | 890 | ||
AB775900.1 | - | 2 | 890 | |||
AB775901.1 | - | 2 | 890 | |||
BC027179.1 | - | 2 | 951 | 3923096 | ||
J03581.1 | - | 2 | 887 | |||
M27160.1 | - | 2 | 1392 | |||
M74314.1 | - | 2 | 927 | |||
U01873.1 | - | 2 | 890 | |||
Y00819.1 | - | 2 | 896 |