Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TYR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000372.4 | NP_000363.1 | 5 | 1647 | ||
GenBank mRNA | AB775899.1 | - | 5 | 1565 | ||
AB775900.1 | - | 5 | 1565 | |||
AB775901.1 | - | 5 | 1565 | |||
M27160.1 | - | 5 | 2067 | |||
M74314.1 | - | 5 | 1611 | |||
U01873.1 | - | 5 | 1565 | |||
Y00819.1 | - | 5 | 1571 |