Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDKN2D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001800.3 | NP_001791.1 | 2 | 921 | ||
NM_079421.2 | NP_524145.1 | 3 | 667 | |||
GenBank mRNA | BC001822.2 | - | 3 | 651 | 3529045 | |
BI669755.1 | - | 3 | 672 | |||
U20498.1 | - | 2 | 620 | |||
U40343.1 | - | 2 | 760 |