Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PECAM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000442.4 | NP_000433.4 | 16 | 2722 | ||
XM_005276880.1 | XP_005276937.1 | 15 | 2985 | |||
XM_005276881.1 | XP_005276938.1 | 15 | 2951 | |||
XM_005276882.1 | XP_005276939.1 | 15 | 2945 | |||
XM_005276883.2 | XP_005276940.1 | 14 | 2922 | |||
XM_011524889.2 | XP_011523191.1 | 17 | 2738 | |||
XM_011524890.1 | XP_011523192.1 | 17 | 2687 | |||
XM_017024738.1 | XP_016880227.1 | 17 | 2631 | |||
XM_017024739.1 | XP_016880228.1 | 15 | 2954 | |||
XM_017024740.1 | XP_016880229.1 | 16 | 2644 | |||
XM_017024741.1 | XP_016880230.1 | 14 | 2897 | |||
GenBank mRNA | BC022512.1 | - | 16 | 2702 | 4816878 | |
BC051822.1 | - | 16 | 2722 | 5768579 |