Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LDLR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000527.4 | NP_000518.1 | 17 | 2700 | ||
NM_001195798.1 | NP_001182727.1 | 17 | 2700 | |||
NM_001195799.1 | NP_001182728.1 | 16 | 2577 | |||
NM_001195800.1 | NP_001182729.1 | 15 | 2196 | |||
NM_001195803.1 | NP_001182732.1 | 15 | 2166 | |||
XM_011528010.1 | XP_011526312.1 | 16 | 2585 | |||
GenBank mRNA | AB209409.1 | - | 17 | 2584 | ||
AK295612.1 | - | 16 | 2236 | |||
AK296312.1 | - | 15 | 2065 | |||
AK299038.1 | - | 15 | 2095 | |||
AK300313.1 | - | 16 | 2476 | |||
AY114155.1 | - | 17 | 2527 | |||
BC014514.1 | - | 17 | 2583 | 3542845 | ||
BT007361.1 | - | 17 | 2513 | |||
BX648281.1 | - | 17 | 2768 | |||
S70123.1 | - | 2 | 264 |