Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDHB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000925.3 | NP_000916.2 | 6 | 417 | ||
NM_001173468.1 | NP_001166939.1 | 6 | 417 | |||
NM_001315536.1 | NP_001302465.1 | 5 | 363 | |||
NR_033384.1 | 5 | 487 | ||||
GenBank mRNA | AB209241.1 | - | 3 | 2779 | ||
AK225569.1 | - | 6 | 392 | |||
AK225621.1 | - | 6 | 397 | |||
AK293153.1 | - | 6 | 383 | |||
AK311514.1 | - | 6, 7 | 392 | |||
AK313022.1 | - | 6 | 392 | |||
AL117618.1 | - | 5 | 328 | |||
BC000439.2 | - | 6 | 384 | 2820672 | ||
BC001924.1 | - | 6 | 384 | 2820672 | ||
CR541911.1 | - | 6 | 374 | |||
DA965753.1 | - | 6 | 417 | |||
DB039833.1 | - | 5 | 487 | |||
J03576.1 | - | 6 | 392 | |||
M34055.1 | - | 6 | 379 | |||
M34056.1 | - | 5 | 302 | |||
M34479.1 | - | 6 | 374 | |||
M54788.1 | - | 6 | 379 | |||
X57778.1 | - | 6 | 393 |