Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: SLC2A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042.2 | NP_001033.1 | 11 | 2076 | ||
GenBank mRNA | BC034387.1 | - | 11 | 2137 | 5187454 | |
M20747.1 | - | 11 | 2021 |