Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other FHL1 SIRNA Assays ›
Gene Symbol:
FHL1
Gene Name:
four and a half LIM domains 1
Gene Aliases:
FHL-1, FHL1A, FHL1B, FLH1A, KYOT, RBMX1A, RBMX1B, SLIM, SLIM-1, SLIM1, SLIMMER, XMPMA
Chromosome Location:
Chr.X: 136146702 - 136211359 on Build GRCh38
UniGene ID:
Hs.435369
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
106450
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: FHL1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001159699.1
NP_001153171.1
6
1077
NM_001159700.1
NP_001153172.1
7
1072
NM_001159701.1
NP_001153173.1
6
948
NM_001159702.2
NP_001153174.1
8
1341
NM_001159703.1
NP_001153175.1
6
954
NM_001159704.1
NP_001153176.1
6
1523
NM_001167819.1
NP_001161291.1
7
1009
NM_001449.4
NP_001440.2
7
1141
NR_027621.1
7
1223
XM_006724743.2
XP_006724806.1
7
1145
XM_006724744.2
XP_006724807.1
8
1223
XM_006724745.3
XP_006724808.1
8
1161
XM_006724746.2
XP_006724809.1
7
1096
XM_006724747.2
XP_006724810.1
6
896
XM_011531316.1
XP_011529618.1
5
758
XM_017029357.1
XP_016884846.1
7
1023
GenBank mRNA
AF063002.1
-
7
1096
AF098518.1
-
7
1065
AK091702.1
-
6
639
AK094024.1
-
5
657
AK122708.1
-
6
1524
AK223299.1
-
5
846
AK289411.1
-
7
1223
AK297124.1
-
7
914
AK299379.1
-
6
803
AK299381.1
-
6
923
AK299450.1
-
7
857
AK300966.1
-
8
948
AK301642.1
-
6
945
AK304680.1
-
7
1000
BC010998.2
-
7
1020
4039973
BC088369.1
-
6
910
30529295
CR456974.1
-
5
812
U29538.1
-
5
833
U60115.1
-
6
895
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
regulation of membrane depolarization
muscle organ development
organ morphogenesis
negative regulation of G2/M transition of mitotic cell cycle
cell differentiation
negative regulation of cell growth
positive regulation of potassium ion transport
regulation of potassium ion transmembrane transporter activity
negative regulation of G1/S transition of mitotic cell cycle
molecular_function
protein binding
zinc ion binding
ion channel binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-d4wl8:80/10.239.162.132:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0