Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FHL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039492.2 | NP_001034581.1 | 7 | 1139 | ||
NM_001318894.1 | NP_001305823.1 | 6 | 1385 | |||
NM_001318895.1 | NP_001305824.1 | 7 | 1373 | |||
NM_001318896.1 | NP_001305825.1 | 6 | 1088 | |||
NM_001318897.1 | NP_001305826.1 | 5 | 1139 | |||
NM_001318898.1 | NP_001305827.1 | 6 | 1198 | |||
NM_001318899.1 | NP_001305828.1 | 8 | 1261 | |||
NM_001450.3 | NP_001441.4 | 6 | 1322 | |||
NM_201555.1 | NP_963849.1 | 8 | 1494 | |||
NM_201557.3 | NP_963851.2 | 6 | 1314 | |||
XM_011510798.2 | XP_011509100.1 | 7 | 1121 | |||
GenBank mRNA | AB158503.1 | - | 5 | 861 | ||
AK097756.1 | - | 1 | 1725 | |||
BC012742.2 | - | 8 | 1494 | 3840517 | ||
BC014397.1 | - | 8 | 1494 | 3546686 | ||
BC036085.1 | - | 6 | 969 | |||
BC093049.1 | - | 6 | 1080 | 30374919 | ||
BI092736.1 | - | 3 | 432 | |||
BX340370.2 | - | 2 | 282 | |||
CR936682.1 | - | 6 | 1385 | |||
CR936799.1 | - | 6 | 6366 | |||
DQ266432.1 | - | 7 | 1373 | |||
L42176.1 | - | 7 | 1053 | |||
U29332.1 | - | 6 | 1036 | |||
U60117.1 | - | 3 | 538 |