Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007240.2 | NP_001007241.2 | 6 | 903 | ||
NM_001007241.2 | NP_001007242.2 | 5 | 462 | |||
NM_001007242.2 | NP_001007243.2 | 4 | 453 | |||
NM_001502.3 | NP_001493.2 | 5 | 894 | |||
XM_017023155.1 | XP_016878644.1 | 5 | 647 | |||
GenBank mRNA | AB035541.1 | - | 4 | 683 | ||
AB035542.1 | - | 3 | 242 | |||
AK293384.1 | - | 5 | 650 | |||
AK312277.1 | - | 5 | 781 | |||
BC032693.2 | - | 3 | 300 | 5229838 | ||
BG661274.1 | - | 3 | 300 | |||
BX647831.1 | - | 6 | 791 | |||
BX648023.1 | - | 5 | 788 | |||
BX648201.1 | - | 4 | 340 | |||
BX649067.1 | - | 5 | 349 | |||
D38225.1 | - | 4 | 254 | |||
U36221.1 | - | 5 | 763 |