Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AMPH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001635.3 | NP_001626.1 | 18 | 1751 | ||
NM_139316.2 | NP_647477.1 | 17 | 1625 | |||
XM_006715689.3 | XP_006715752.1 | 17 | 3288 | |||
XM_006715690.3 | XP_006715753.1 | 17 | 2070 | |||
XM_011515271.2 | XP_011513573.1 | 18 | 2196 | |||
XM_017011995.1 | XP_016867484.1 | 18 | 3414 | |||
XM_017011996.1 | XP_016867485.1 | 17 | 2376 | |||
GenBank mRNA | AF034996.1 | - | 17 | 1478 | ||
AF498092.1 | - | 6 | 806 | |||
AF498093.1 | - | 4 | 763 | |||
AF498094.1 | - | 4 | 1675 | |||
AF498095.1 | - | 5 | 562 | |||
AF498096.1 | - | 9 | 1201 | |||
AK291447.1 | - | 18 | 1555 | |||
BC034376.1 | - | 18 | 1542 | 5181100 | ||
U07616.1 | - | 18 | 1645 | |||
X81438.1 | - | 18 | 1609 |