Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: CPB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278541.1 | NP_001265470.1 | 5, 6 | 561 | ||
NM_001872.4 | NP_001863.3 | 5, 6 | 561 | |||
XM_017020393.1 | XP_016875882.1 | 5, 6 | 504 | |||
GenBank mRNA | AB011969.1 | - | 5, 6 | 513 | ||
AK290829.1 | - | 5, 6 | 509 | |||
BC007057.1 | - | 5, 6 | 518 | 3934520 | ||
BT006936.1 | - | 5, 6 | 494 | |||
M75106.1 | - | 5, 6 | 513 |