Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD1D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK225996.1 | - | 5 | 1000 | ||
BC027926.1 | - | 7 | 1565 | 5229534 | ||
J04142.1 | - | 6 | 1230 |