Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256030.1 | NP_001242959.1 | 2 | 347 | ||
NM_001778.3 | NP_001769.2 | 2 | 347 | |||
XM_005245625.1 | XP_005245682.1 | 2 | 318 | |||
XM_011510171.1 | XP_011508473.1 | 3 | 415 | |||
GenBank mRNA | AK225636.1 | - | 2 | 320 | ||
AK310624.1 | - | 2 | 321 | |||
AK310744.1 | - | 2 | 327 | |||
BC016182.1 | - | 2 | 291 | 4850933 | ||
BC030224.1 | - | 2 | 299 | 5223308 | ||
BG505217.1 | - | 2 | 347 | |||
BT019813.1 | - | 2 | 259 | |||
CR457012.1 | - | 2 | 259 | |||
M37766.1 | - | 2 | 279 | |||
M59904.1 | - | 2 | 276 | |||
X06341.1 | - | 2 | 295 |