Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRHBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001882.3 | NP_001873.2 | 3, 4 | 621 | ||
GenBank mRNA | AK223457.1 | - | 3, 4 | 498 | ||
AK307729.1 | - | 3, 4 | 431 | |||
AK311052.1 | - | 3, 4 | 427 | |||
BC018038.1 | - | 3, 4 | 431 | 4812893 | ||
CB959505.1 | - | 3, 4 | 648 | |||
CR541666.1 | - | 3, 4 | 341 | |||
X58022.1 | - | 3, 4 | 387 |