Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DLAT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001931.4 | NP_001922.2 | 7 | 1723 | ||
XM_011542647.2 | XP_011540949.1 | 7 | 1370 | |||
GenBank mRNA | AK057299.1 | - | 7 | 1723 | ||
AK223596.1 | - | 7 | 1112 | |||
AK296274.1 | - | 7 | 1002 | |||
AK299562.1 | - | 5 | 560 | |||
AK303930.1 | - | 8 | 948 | |||
AL832703.1 | - | 7 | 1094 | |||
BC039084.1 | - | 7 | 1092 | 4508489 | ||
X13822.1 | - | 1 | 56 | |||
Y00978.1 | - | 7 | 968 |