Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HIVEP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002114.3 | NP_002105.3 | 4 | 5632 | ||
XM_011514546.2 | XP_011512848.1 | 6 | 6675 | |||
XM_011514547.2 | XP_011512849.1 | 5 | 5834 | |||
XM_011514548.2 | XP_011512850.1 | 5 | 6078 | |||
XM_011514549.2 | XP_011512851.1 | 5 | 5958 | |||
XM_011514550.2 | XP_011512852.1 | 5 | 5825 | |||
XM_011514551.2 | XP_011512853.1 | 5 | 6503 | |||
XM_011514552.2 | XP_011512854.1 | 4 | 5663 | |||
XM_011514553.1 | XP_011512855.1 | 4 | 5644 | |||
XM_011514555.2 | XP_011512857.1 | 3 | 7233 | |||
XM_017010800.1 | XP_016866289.1 | 7 | 6561 | |||
XM_017010801.1 | XP_016866290.1 | 7 | 6502 | |||
XM_017010802.1 | XP_016866291.1 | 5 | 6581 | |||
XM_017010803.1 | XP_016866292.1 | 6 | 6676 | |||
XM_017010804.1 | XP_016866293.1 | 6 | 6679 | |||
GenBank mRNA | BC140816.1 | - | 3 | 5565 | 9021684 | |
M32019.1 | - | 1 | 3005 | |||
X51435.1 | - | 4 | 5774 |