Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ITPR1 SIRNA Assays ›
Gene Symbol:
ITPR1
Gene Name:
inositol 1,4,5-trisphosphate receptor type 1
Gene Aliases:
ACV, CLA4, INSP3R1, IP3R, IP3R1, PPP1R94, SCA15, SCA16, SCA29
Chromosome Location:
Chr.3: 4493348 - 4847840 on Build GRCh38
UniGene ID:
Hs.567295
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
106734
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ITPR1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001099952.2
NP_001093422.2
15, 16
1770
NM_001168272.1
NP_001161744.1
14, 15
1725
NM_002222.5
NP_002213.5
14, 15
1725
XM_005265109.2
XP_005265166.1
15, 16
1770
XM_005265110.2
XP_005265167.1
15, 16
1770
XM_006713131.2
XP_006713194.1
15, 16
1770
XM_011533681.1
XP_011531983.1
15, 16
1770
XM_011533682.2
XP_011531984.1
15, 16
1696
XM_011533683.2
XP_011531985.1
15, 16
1770
XM_011533684.1
XP_011531986.1
15, 16
1770
XM_011533685.1
XP_011531987.1
15, 16
1770
XM_011533686.1
XP_011531988.1
15, 16
1770
XM_011533687.1
XP_011531989.1
14, 15
1725
XM_011533688.1
XP_011531990.1
14, 15
1725
XM_011533690.1
XP_011531992.1
15, 16
1770
XM_011533691.1
XP_011531993.1
15, 16
1770
XM_011533692.2
XP_011531994.1
15, 16
1770
XM_017006357.1
XP_016861846.1
15, 16
1770
XM_017006358.1
XP_016861847.1
15, 16
1770
GenBank mRNA
AK293752.1
-
9, 10
956
D26070.1
-
14, 15
1631
L38019.2
-
15, 16
1667
U23850.1
-
12, 13
1375
Back To Top
More Information
Panther Classification:
Molecular Function -
ligand-gated ion channel
Gene Ontology Categories:
Function(s)
Process(es)
response to hypoxia
calcium ion transport
signal transduction
post-embryonic development
regulation of autophagy
platelet activation
endoplasmic reticulum calcium ion homeostasis
epithelial fluid transport
inositol phosphate-mediated signaling
regulation of insulin secretion
negative regulation of calcium-mediated signaling
voluntary musculoskeletal movement
release of sequestered calcium ion into cytosol
intrinsic apoptotic signaling pathway in response to endoplasmic reticulum stress
regulation of cardiac conduction
inositol 1,4,5-trisphosphate-sensitive calcium-release channel activity
protein binding
calcium ion transmembrane transporter activity
calcium-release channel activity
calcium channel inhibitor activity
phosphatidylinositol binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-6d74b6fc5b-sshrn:80/10.239.165.44:80.
git-commit: 0683b6a9b3dadc77a268b704485bdfd9f1fd09b0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.17.0-2024.10.41-1.0