Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: IPO5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002271.4 | NP_002262.3 | 14 | 1425 | ||
XM_005254052.3 | XP_005254109.1 | 13 | 1335 | |||
XM_005254053.3 | XP_005254110.1 | 13 | 1352 | |||
XM_011521087.2 | XP_011519389.1 | 15 | 1516 | |||
XM_011521088.2 | XP_011519390.1 | 15 | 1582 | |||
XM_011521089.2 | XP_011519391.1 | 16 | 1639 | |||
XM_011521090.2 | XP_011519392.1 | 14 | 1475 | |||
XM_017020562.1 | XP_016876051.1 | 14 | 1456 | |||
GenBank mRNA | AK125031.1 | - | 1 | 904 | ||
AK225286.1 | - | 11 | 1210 | |||
AK302812.1 | - | 11 | 1197 | |||
AK303494.1 | - | 9 | 838 | |||
BC001497.2 | - | 11 | 1193 | 2823581 | ||
BC019309.1 | - | 11 | 1193 | |||
BC045640.1 | - | 14 | 1406 | 5298410 | ||
BX648222.1 | - | 11 | 1255 | |||
U72761.1 | - | 11 | 1191 | |||
Y08890.1 | - | 14 | 1427 |