Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AFF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025108.1 | NP_001020279.1 | 14 | 2518 | ||
NM_002285.2 | NP_002276.2 | 14 | 2535 | |||
XM_005263943.3 | XP_005264000.2 | 13 | 2412 | |||
XM_005263945.3 | XP_005264002.1 | 5 | 1769 | |||
XM_011511169.2 | XP_011509471.1 | 14 | 2515 | |||
XM_011511170.2 | XP_011509472.1 | 15 | 2654 | |||
XM_011511171.2 | XP_011509473.1 | 14 | 2518 | |||
XM_011511172.2 | XP_011509474.1 | 13 | 2470 | |||
XM_011511173.2 | XP_011509475.1 | 14 | 2531 | |||
XM_011511174.2 | XP_011509476.1 | 14 | 2475 | |||
XM_011511175.2 | XP_011509477.1 | 16 | 2765 | |||
XM_011511176.2 | XP_011509478.1 | 13 | 2456 | |||
XM_011511177.2 | XP_011509479.1 | 14 | 2546 | |||
XM_011511178.2 | XP_011509480.1 | 4 | 1277 | |||
XM_011511179.2 | XP_011509481.1 | 4 | 1274 | |||
XM_017004085.1 | XP_016859574.1 | 5 | 1766 | |||
XM_017004086.1 | XP_016859575.1 | 5 | 2247 | |||
XM_017004087.1 | XP_016859576.1 | 5 | 2244 | |||
GenBank mRNA | AB209634.1 | - | 1 | 237 | ||
AK092327.1 | - | 15 | 2607 | |||
AK290468.1 | - | 13 | 2393 | |||
AK297407.1 | - | 15 | 2783 | |||
BC136579.1 | - | 12 | 2318 | 9020569 | ||
BC144266.1 | - | 13 | 2393 | 9052786 | ||
U34360.1 | - | 12 | 2353 |