Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: MPI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289155.1 | NP_001276084.1 | 3 | 332 | ||
NM_001289156.1 | NP_001276085.1 | 2 | 204 | |||
NM_001289157.1 | NP_001276086.1 | 3 | 332 | |||
NM_002435.2 | NP_002426.1 | 3 | 332 | |||
XM_011521592.1 | XP_011519894.1 | 3 | 408 | |||
XM_011521593.2 | XP_011519895.1 | 3 | 390 | |||
XM_017022207.1 | XP_016877696.1 | 3 | 412 | |||
XM_017022208.1 | XP_016877697.1 | 3 | 390 | |||
XM_017022209.1 | XP_016877698.1 | 2 | 179 | |||
GenBank mRNA | AF504648.1 | - | 3 | 297 | ||
AK225149.1 | - | 3 | 295 | |||
AK292374.1 | - | 3 | 285 | |||
AK294030.1 | - | 3 | 301 | |||
AK302354.1 | - | 2 | 173 | |||
BC017351.2 | - | 3 | 281 | 5013257 | ||
BC046357.1 | - | 3 | 287 | 5745956 | ||
BX648475.1 | - | 3 | 309 | |||
CD706167.1 | - | 3 | 323 | |||
X76057.1 | - | 3 | 274 |