Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARID4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002892.3 | NP_002883.3 | 24 | 4600 | ||
NM_023000.2 | NP_075376.2 | 24 | 4438 | |||
NM_023001.2 | NP_075377.2 | 23 | 4393 | |||
XM_005267964.2 | XP_005268021.1 | 24 | 4779 | |||
XM_005267965.2 | XP_005268022.1 | 23 | 4572 | |||
XM_017021560.1 | XP_016877049.1 | 24 | 4800 | |||
XM_017021561.1 | XP_016877050.1 | 24 | 4577 | |||
XM_017021562.1 | XP_016877051.1 | 24 | 4740 | |||
XM_017021563.1 | XP_016877052.1 | 23 | 4593 | |||
XM_017021564.1 | XP_016877053.1 | 22 | 4408 | |||
XM_017021565.1 | XP_016877054.1 | 14 | 4762 | |||
GenBank mRNA | S66427.1 | - | 24 | 4460 |