Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEC14L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039573.2 | NP_001034662.2 | 12 | 1644 | ||
NM_001143998.1 | NP_001137470.1 | 12 | 1644 | |||
NM_001143999.1 | NP_001137471.1 | 12 | 1640 | |||
NM_001144001.1 | NP_001137473.1 | 10 | 1432 | |||
NM_001204408.1 | NP_001191337.1 | 14 | 1989 | |||
NM_001204410.1 | NP_001191339.1 | 13 | 1682 | |||
NM_003003.3 | NP_002994.3 | 12 | 1591 | |||
GenBank mRNA | AB208811.1 | - | 12 | 1623 | ||
AK290913.1 | - | 12 | 1537 | |||
AK293206.1 | - | 10 | 1432 | |||
BC136523.1 | - | 12 | 1479 | 9020513 | ||
BC136525.1 | - | 12 | 1479 | 9020515 | ||
BC142979.1 | - | 12 | 1535 | 8860414 | ||
BX537448.1 | - | 12 | 1545 | |||
D67029.1 | - | 12 | 1627 | |||
FM995495.1 | - | 14 | 1883 | |||
FM995496.1 | - | 13 | 1680 | |||
FM995497.1 | - | 13 | 1779 | |||
U79284.1 | - | 1 | 60 |
Gene Symbol: SEC14L1P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026952.1 | 1 | 1451 |