Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEC14L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039573.2 | NP_001034662.2 | 13 | 1701 | ||
NM_001143998.1 | NP_001137470.1 | 13 | 1701 | |||
NM_001143999.1 | NP_001137471.1 | 13 | 1697 | |||
NM_001144001.1 | NP_001137473.1 | 11 | 1489 | |||
NM_001204408.1 | NP_001191337.1 | 15 | 2046 | |||
NM_001204410.1 | NP_001191339.1 | 14 | 1739 | |||
NM_003003.3 | NP_002994.3 | 13 | 1648 | |||
GenBank mRNA | AB208811.1 | - | 13 | 1680 | ||
AK290913.1 | - | 13 | 1594 | |||
AK293206.1 | - | 11 | 1489 | |||
AK303328.1 | - | 1 | 103 | |||
BC136523.1 | - | 13 | 1536 | 9020513 | ||
BC136525.1 | - | 13 | 1536 | 9020515 | ||
BC142979.1 | - | 13 | 1592 | 8860414 | ||
BX537448.1 | - | 13 | 1602 | |||
D67029.1 | - | 13 | 1684 | |||
FM995495.1 | - | 15 | 1940 | |||
FM995496.1 | - | 14 | 1737 | |||
U79284.1 | - | 2 | 117 |