Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC6A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003042.3 | NP_003033.3 | 16 | 3130 | ||
XM_005265410.4 | XP_005265467.1 | 16 | 4487 | |||
XM_005265411.4 | XP_005265468.1 | 16 | 4426 | |||
XM_006713306.3 | XP_006713369.1 | 16 | 3085 | |||
XM_011534025.2 | XP_011532327.1 | 16 | 5177 | |||
XM_011534027.2 | XP_011532329.1 | 16 | 5116 | |||
XM_017007071.1 | XP_016862560.1 | 17 | 4909 | |||
XM_017007072.1 | XP_016862561.1 | 17 | 4219 | |||
GenBank mRNA | AK090958.1 | - | 1 | 631 | ||
BC033904.1 | - | 16 | 3098 | 5264672 | ||
BC057291.1 | - | 16 | 3109 |