Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC7A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003045.4 | NP_003036.1 | 13 | 3477 | ||
XM_005266507.3 | XP_005266564.1 | 13 | 3272 | |||
XM_017020713.1 | XP_016876202.1 | 13 | 3544 | |||
XM_017020714.1 | XP_016876203.1 | 13 | 3483 | |||
XM_017020715.1 | XP_016876204.1 | 13 | 3365 | |||
XM_017020716.1 | XP_016876205.1 | 13 | 3488 | |||
GenBank mRNA | AB105377.1 | - | 13 | 3364 | ||
AK124591.1 | - | 1 | 798 | |||
BC063303.1 | - | 13 | 3274 | 30340706 | ||
BM458630.1 | - | 1 | 764 |