Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SORL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003105.5 | NP_003096.1 | 15 | 2228 | ||
XM_011542963.2 | XP_011541265.1 | 15 | 2103 | |||
XM_011542965.2 | XP_011541267.1 | 9 | 1137 | |||
XM_017018169.1 | XP_016873658.1 | 15 | 1935 | |||
XM_017018170.1 | XP_016873659.1 | 13 | 1611 | |||
XM_017018171.1 | XP_016873660.1 | 15 | 2103 | |||
XM_017018172.1 | XP_016873661.1 | 2 | 192 | |||
GenBank mRNA | BC137171.1 | - | 15 | 2151 | 9021168 | |
Y08110.1 | - | 15 | 2179 |