Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TCF4 SIRNA Assays ›
Gene Symbol:
TCF4
Gene Name:
transcription factor 4
Gene Aliases:
E2-2, FECD3, ITF-2, ITF2, PTHS, SEF-2, SEF2, SEF2-1, SEF2-1A, SEF2-1B, SEF2-1D, TCF-4, bHLHb19
Chromosome Location:
Chr.18: 55222331 - 55635993 on Build GRCh38
UniGene ID:
Hs.605153
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
107027
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TCF4
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001083962.1
NP_001077431.1
14
1749
NM_001243226.2
NP_001230155.2
15
1539
NM_001243227.1
NP_001230156.1
13
1583
NM_001243228.1
NP_001230157.1
14
1693
NM_001243230.1
NP_001230159.1
13
1222
NM_001243231.1
NP_001230160.1
12
1183
NM_001243232.1
NP_001230161.1
10
1205
NM_001243233.1
NP_001230162.1
10
1146
NM_001243234.1
NP_001230163.1
7
778
NM_001243235.1
NP_001230164.1
7
778
NM_001243236.1
NP_001230165.1
7
794
NM_001306207.1
NP_001293136.1
13
1374
NM_001306208.1
NP_001293137.1
10
1205
NM_003199.2
NP_003190.1
14
1749
XM_005266739.3
XP_005266796.2
13
1389
XM_005266741.3
XP_005266798.1
14
1349
XM_005266743.4
XP_005266800.1
13
1210
XM_005266744.3
XP_005266801.1
13
1330
XM_005266745.3
XP_005266802.1
13
1192
XM_005266747.3
XP_005266804.1
12
1186
XM_005266749.3
XP_005266806.1
10
977
XM_005266752.4
XP_005266809.1
10
1547
XM_005266754.3
XP_005266811.1
9
971
XM_005266755.4
XP_005266812.1
10
1006
XM_005266761.3
XP_005266818.1
7
1287
XM_006722536.2
XP_006722599.1
14
1672
XM_006722537.2
XP_006722600.1
14
1305
XM_006722538.2
XP_006722601.1
13
1327
XM_011526158.2
XP_011524460.1
13
2303
XM_011526160.2
XP_011524462.1
9
1356
XM_017025934.1
XP_016881423.1
13
1386
XM_017025935.1
XP_016881424.1
13
1386
XM_017025936.1
XP_016881425.1
13
1389
XM_017025937.1
XP_016881426.1
13
1143
XM_017025938.1
XP_016881427.1
14
1302
XM_017025939.1
XP_016881428.1
14
1349
XM_017025940.1
XP_016881429.1
14
1672
XM_017025941.1
XP_016881430.1
14
1305
XM_017025942.1
XP_016881431.1
14
1349
XM_017025943.1
XP_016881432.1
14
1349
XM_017025944.1
XP_016881433.1
13
1607
XM_017025945.1
XP_016881434.1
13
1607
XM_017025946.1
XP_016881435.1
13
1624
XM_017025947.1
XP_016881436.1
13
1210
XM_017025948.1
XP_016881437.1
13
1607
XM_017025949.1
XP_016881438.1
13
1327
XM_017025950.1
XP_016881439.1
13
1604
XM_017025951.1
XP_016881440.1
12
1183
XM_017025952.1
XP_016881441.1
10
1197
XM_017025953.1
XP_016881442.1
10
977
XM_017025954.1
XP_016881443.1
10
1558
XM_017025955.1
XP_016881444.1
9
1292
XM_017025956.1
XP_016881445.1
9
971
XM_017025957.1
XP_016881446.1
7
1287
GenBank mRNA
AB209741.1
-
9
857
AK095041.1
-
10
1205
AK096862.1
-
12
1183
AK122765.1
-
14
1749
AK299169.1
-
13
1583
AK300038.1
-
7
724
AK300612.1
-
10
1146
AK300636.1
-
9
876
AK301144.1
-
13
1360
AK315074.1
-
15
1681
AK316165.1
-
9
920
AL518903.3
-
7
794
BC031056.1
-
15
1500
BC125084.1
-
13
1163
40117795
BC125085.1
-
13
1163
40117798
FR748210.1
-
13
1137
FR748211.1
-
13
1137
FR748212.1
-
11
957
FR748213.1
-
11
957
FR748214.1
-
12
1065
FR748215.1
-
10
885
FR748216.1
-
13
1128
FR748217.1
-
12
1011
FR748218.1
-
8
747
FR748219.1
-
10
924
FR748220.1
-
7
657
FR748221.1
-
7
657
FR748222.1
-
7
657
FR748223.1
-
5
489
M74718.1
-
7
609
M74719.1
-
14
1336
X52079.1
-
11
1002
Back To Top
More Information
Panther Classification:
Molecular Function -
basic helix-loop-helix transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
DNA-templated transcription, initiation
transcription initiation from RNA polymerase II promoter
positive regulation of neuron differentiation
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
protein-DNA complex assembly
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcription factor activity, sequence-specific DNA binding, RNA polymerase recruiting
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
transcription factor activity, TFIIB-class binding
TFIIB-class transcription factor binding
DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
protein C-terminus binding
identical protein binding
protein homodimerization activity
bHLH transcription factor binding
protein heterodimerization activity
E-box binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-648b49bbc4-xm8cg:80/10.239.167.213:80.
git-commit: cd89e1917689fc5870ae005717bc737506e58003
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.17.2-2024.10.99-1.0