Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TCF4 SIRNA Assays ›
Gene Symbol:
TCF4
Gene Name:
transcription factor 4
Gene Aliases:
E2-2, FECD3, ITF-2, ITF2, PTHS, SEF-2, SEF2, SEF2-1, SEF2-1A, SEF2-1B, SEF2-1D, TCF-4, bHLHb19
Chromosome Location:
Chr.18: 55222331 - 55635993 on Build GRCh38
UniGene ID:
Hs.605153
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
107028
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TCF4
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001083962.1
NP_001077431.1
19
2503
NM_001243226.2
NP_001230155.2
20
2293
NM_001243227.1
NP_001230156.1
18
2337
NM_001243228.1
NP_001230157.1
19
2447
NM_001243230.1
NP_001230159.1
18
1964
NM_001243231.1
NP_001230160.1
17
1925
NM_001243232.1
NP_001230161.1
15
1959
NM_001243233.1
NP_001230162.1
15
1888
NM_001243234.1
NP_001230163.1
12
1532
NM_001243235.1
NP_001230164.1
12
1520
NM_001243236.1
NP_001230165.1
12
1536
NM_001306207.1
NP_001293136.1
18
2116
NM_001306208.1
NP_001293137.1
15
1947
NM_003199.2
NP_003190.1
19
2491
XM_005266739.3
XP_005266796.2
18
2143
XM_005266741.3
XP_005266798.1
19
2103
XM_005266743.4
XP_005266800.1
18
1964
XM_005266744.3
XP_005266801.1
18
2084
XM_005266745.3
XP_005266802.1
18
1946
XM_005266747.3
XP_005266804.1
17
1940
XM_005266749.3
XP_005266806.1
15
1731
XM_005266752.4
XP_005266809.1
15
2301
XM_005266754.3
XP_005266811.1
14
1725
XM_005266755.4
XP_005266812.1
15
1760
XM_005266761.3
XP_005266818.1
12
2041
XM_006722536.2
XP_006722599.1
19
2426
XM_006722537.2
XP_006722600.1
19
2059
XM_006722538.2
XP_006722601.1
18
2081
XM_011526158.2
XP_011524460.1
18
3057
XM_011526160.2
XP_011524462.1
14
2110
XM_017025934.1
XP_016881423.1
18
2140
XM_017025935.1
XP_016881424.1
18
2140
XM_017025936.1
XP_016881425.1
18
2131
XM_017025937.1
XP_016881426.1
18
1885
XM_017025938.1
XP_016881427.1
19
2056
XM_017025939.1
XP_016881428.1
19
2103
XM_017025940.1
XP_016881429.1
19
2414
XM_017025941.1
XP_016881430.1
19
2047
XM_017025942.1
XP_016881431.1
19
2091
XM_017025943.1
XP_016881432.1
19
2091
XM_017025944.1
XP_016881433.1
18
2361
XM_017025945.1
XP_016881434.1
18
2361
XM_017025946.1
XP_016881435.1
18
2366
XM_017025947.1
XP_016881436.1
18
1952
XM_017025948.1
XP_016881437.1
18
2349
XM_017025949.1
XP_016881438.1
18
2069
XM_017025950.1
XP_016881439.1
18
2346
XM_017025951.1
XP_016881440.1
17
1937
XM_017025952.1
XP_016881441.1
15
1951
XM_017025953.1
XP_016881442.1
15
1719
XM_017025954.1
XP_016881443.1
15
2312
XM_017025955.1
XP_016881444.1
14
2046
XM_017025956.1
XP_016881445.1
14
1713
XM_017025957.1
XP_016881446.1
12
2029
GenBank mRNA
AB209741.1
-
13
5041
AK026674.1
-
2
1634
AK095041.1
-
15
1959
AK096862.1
-
17
1925
AK122765.1
-
19
2491
AK299169.1
-
18
2337
AK300038.1
-
12
1478
AK300612.1
-
15
1888
AK300636.1
-
14
1618
AK301144.1
-
18
2102
AK315074.1
-
20
2435
AK316165.1
-
14
1662
BC031056.1
-
20
2254
BC125084.1
-
18
1917
40117795
BC125085.1
-
18
1905
40117798
FR748210.1
-
18
1891
FR748211.1
-
18
1879
FR748212.1
-
16
1711
FR748213.1
-
16
1699
FR748214.1
-
17
1807
FR748215.1
-
15
1627
FR748216.1
-
18
1870
FR748217.1
-
17
1753
FR748218.1
-
13
1489
FR748219.1
-
15
1666
FR748220.1
-
12
1411
FR748221.1
-
12
1399
FR748222.1
-
12
1399
FR748223.1
-
10
1231
M74718.1
-
12
1363
M74719.1
-
19
2078
X52079.1
-
16
1744
Back To Top
More Information
Panther Classification:
Molecular Function -
basic helix-loop-helix transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
DNA-templated transcription, initiation
transcription initiation from RNA polymerase II promoter
positive regulation of neuron differentiation
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
protein-DNA complex assembly
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcription factor activity, sequence-specific DNA binding, RNA polymerase recruiting
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
transcription factor activity, TFIIB-class binding
TFIIB-class transcription factor binding
DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
protein C-terminus binding
identical protein binding
protein homodimerization activity
bHLH transcription factor binding
protein heterodimerization activity
E-box binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-648b49bbc4-xm8cg:80/10.239.167.213:80.
git-commit: cd89e1917689fc5870ae005717bc737506e58003
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.17.2-2024.10.99-1.0