Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TCF12 SIRNA Assays ›
Gene Symbol:
TCF12
Gene Name:
transcription factor 12
Gene Aliases:
CRS3, HEB, HTF4, HsT17266, TCF-12, bHLHb20
Chromosome Location:
Chr.15: 56918090 - 57291261 on Build GRCh38
UniGene ID:
Hs.511504
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
107035
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TCF12
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001306219.2
NP_001293148.1
14
2824
NM_001306220.2
NP_001293149.1
12
2612
NM_001322151.1
NP_001309080.1
21
3531
NM_001322152.1
NP_001309081.1
21
3533
NM_001322154.1
NP_001309083.1
21
3531
NM_001322156.1
NP_001309085.1
22
3598
NM_001322158.1
NP_001309087.1
21
3526
NM_001322161.1
NP_001309090.1
21
3379
NM_001322162.1
NP_001309091.1
21
3382
NM_001322164.1
NP_001309093.1
20
3329
NM_001322165.1
NP_001309094.1
19
3293
NM_003205.3
NP_003196.1
20
3464
NM_207036.1
NP_996919.1
21
3526
NM_207037.1
NP_996920.1
21
3536
NM_207038.1
NP_996921.1
20
3454
NM_207040.1
NP_996923.1
13
2725
XM_011521959.2
XP_011520261.1
22
3532
XM_011521961.2
XP_011520263.1
22
3529
XM_011521962.2
XP_011520264.1
21
3460
XM_011521963.2
XP_011520265.1
21
3457
XM_011521965.1
XP_011520267.1
20
3243
XM_011521966.2
XP_011520268.1
13
3139
XM_011521967.1
XP_011520269.1
19
3171
XM_011521969.1
XP_011520271.1
13
2715
XM_017022520.1
XP_016878009.1
20
3427
GenBank mRNA
AL831981.2
-
20
3432
BC050556.1
-
21
3477
5767579
BX537967.1
-
13
2715
M65209.1
-
8
2117
M80627.1
-
20
3383
M83233.1
-
20
3258
Back To Top
More Information
Panther Classification:
Molecular Function -
basic helix-loop-helix transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
immune response
muscle organ development
positive regulation of neuron differentiation
positive regulation of transcription from RNA polymerase II promoter
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
transcription factor activity, sequence-specific DNA binding
protein binding
transcription factor binding
enhancer binding
cAMP response element binding
protein homodimerization activity
bHLH transcription factor binding
transcription regulatory region DNA binding
SMAD binding
protein heterodimerization activity
E-box binding
HMG box domain binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-qqtbl:80/10.239.164.61:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0