Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TUB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003320.4 | NP_003311.2 | 8 | 1197 | ||
NM_177972.2 | NP_813977.1 | 7 | 940 | |||
XM_005253109.3 | XP_005253166.1 | 7 | 932 | |||
XM_011520344.2 | XP_011518646.1 | 7 | 987 | |||
GenBank mRNA | AB209618.1 | - | 1 | 4395 | ||
AK293528.1 | - | 7 | 868 | |||
BC075031.2 | - | 8 | 989 | 30915625 | ||
BC075032.2 | - | 8 | 989 | 30915418 | ||
U54644.1 | - | 7 | 943 | |||
U82467.1 | - | 8 | 1197 |