Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC4A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098484.2 | NP_001091954.1 | 6 | 734 | ||
NM_001134742.1 | NP_001128214.1 | 6 | 734 | |||
NM_003759.3 | NP_003750.1 | 3 | 681 | |||
XM_017008792.1 | XP_016864281.1 | 4 | 407 | |||
GenBank mRNA | AB470072.1 | - | 5 | 624 | ||
AF004813.1 | - | 4 | 557 | |||
AF007216.1 | - | 3 | 634 | |||
AF011390.1 | - | 6 | 734 | |||
AF053753.1 | - | 5 | 617 | |||
AF053754.1 | - | 5 | 617 | |||
AF069510.1 | - | 5 | 661 | |||
AF157492.1 | - | 6 | 693 | |||
AF310248.1 | - | 5 | 617 | |||
AK226164.1 | - | 4 | 507 | |||
AK307594.1 | - | 6 | 714 | |||
AK308038.1 | - | 4 | 208 | |||
BC030977.1 | - | 3 | 681 | 4610968 | ||
BC127908.1 | - | 3 | 278 | |||
EF531618.1 | - | 5 | 617 |