Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MPDZ SIRNA Assays ›
Gene Symbol:
MPDZ
Gene Name:
multiple PDZ domain crumbs cell polarity complex component
Gene Aliases:
HYC2, MUPP1
Chromosome Location:
Chr.9: 13105296 - 13279687 on Build GRCh38
UniGene ID:
Hs.169378
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
107246
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MPDZ
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001261406.1
NP_001248335.1
37
5334
NM_001261407.1
NP_001248336.1
37
5334
NM_003829.4
NP_003820.2
38
5433
XM_005251622.4
XP_005251679.1
38
5382
XM_005251623.4
XP_005251680.1
36
5172
XM_006716885.3
XP_006716948.1
38
5431
XM_006716886.3
XP_006716949.1
39
5590
XM_006716887.3
XP_006716950.1
38
5534
XM_006716888.3
XP_006716951.1
38
5382
XM_006716889.3
XP_006716952.1
37
5286
XM_006716891.3
XP_006716954.1
37
5286
XM_017015252.1
XP_016870741.1
37
5298
XM_017015253.1
XP_016870742.1
37
5271
XM_017015254.1
XP_016870743.1
37
5383
XM_017015255.1
XP_016870744.1
36
5231
XM_017015256.1
XP_016870745.1
36
5172
XM_017015257.1
XP_016870746.1
36
5159
GenBank mRNA
AB210041.1
-
38
5434
AF093419.1
-
37
5257
AJ001319.1
-
3
360
AK074721.1
-
7
835
AK091945.1
-
13
1711
AK098775.1
-
5
611
AK294823.1
-
11
1343
AK294976.1
-
16
1939
AK307924.1
-
11
1342
AK307988.1
-
14
1751
AK316489.1
-
15
1817
BC140793.1
-
36
5158
9021661
BC144564.1
-
36
5158
9053092
CR936648.1
-
22
5321
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
cell adhesion
viral process
myelination
protein binding
protein C-terminus binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-xql29:80/10.239.165.153:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0