Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003852.3 | NP_003843.3 | 5 | 1085 | ||
NM_015905.2 | NP_056989.2 | 5 | 1085 | |||
GenBank mRNA | AF009353.1 | - | 5 | 858 | ||
AF119042.1 | - | 5 | 986 | |||
AK075306.1 | - | 5 | 1085 | |||
AK302679.1 | - | 6 | 958 | |||
AK312610.1 | - | 5 | 1213 | |||
BC028689.2 | - | 5 | 983 | 4827559 |