Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACOX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001185039.1 | NP_001171968.1 | 8 | 1590 | ||
NM_004035.6 | NP_004026.2 | 8 | 1416 | |||
NM_007292.5 | NP_009223.2 | 8 | 1416 | |||
XM_011524868.2 | XP_011523170.1 | 9 | 1575 | |||
XM_011524869.2 | XP_011523171.1 | 7 | 1253 | |||
GenBank mRNA | AK225897.1 | - | 8 | 1116 | ||
AK291793.1 | - | 8 | 1110 | |||
AK292965.1 | - | 8 | 1115 | |||
AK296409.1 | - | 8 | 1230 | |||
AK300447.1 | - | 9 | 1271 | |||
AK308218.1 | - | 7 | 855 | |||
BC008767.2 | - | 8 | 1345 | 3051501 | ||
BC010425.1 | - | 8 | 1134 | 3357409 | ||
BQ943492.1 | - | 5 | 625 | |||
BX537380.1 | - | 8 | 1075 | |||
S69189.1 | - | 8 | 1107 | |||
U07866.1 | - | 8 | 1055 | |||
X71440.1 | - | 8 | 1068 |
Gene Symbol: GJA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK310985.1 | - | 776 |