Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ECHS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004092.3 | NP_004083.3 | 5, 6 | 698 | ||
GenBank mRNA | BC008906.2 | - | 5, 6 | 641 | 2907059 | |
BC013264.1 | - | 5, 6 | 633 | |||
BQ130816.1 | - | 3, 4 | 333 | |||
BT007123.1 | - | 5, 6 | 627 | |||
D13900.1 | - | 5, 6 | 648 |