Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC16A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042422.2 | NP_001035887.1 | 5 | 1771 | ||
NM_001042423.2 | NP_001035888.1 | 5 | 1775 | |||
NM_001206950.1 | NP_001193879.1 | 5 | 1787 | |||
NM_001206951.1 | NP_001193880.1 | 5 | 1749 | |||
NM_001206952.1 | NP_001193881.1 | 5 | 1716 | |||
NM_004207.3 | NP_004198.1 | 5 | 1743 | |||
XM_011523607.1 | XP_011521909.1 | 5 | 3664 | |||
GenBank mRNA | AK127319.1 | - | 4 | 3931 | ||
AK223040.1 | - | 5 | 1701 | |||
U81800.1 | - | 5 | 1683 |