Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006658.2 | NP_001006659.1 | 16 | 3087 | ||
NM_001877.4 | NP_001868.2 | 15 | 2910 | |||
XM_011509206.2 | XP_011507508.1 | 16 | 2983 | |||
GenBank mRNA | AK223627.1 | - | 16 | 3059 | ||
AK301496.1 | - | 16 | 3055 | |||
BC090937.1 | - | 15 | 2845 | 3066233 | ||
BC136394.1 | - | 16 | 3061 | 9020381 | ||
J03565.1 | - | 16 | 3016 | |||
M26004.1 | - | 15 | 2860 | |||
Y00649.1 | - | 15 | 2843 |