Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOTCH4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004557.3 | NP_004548.3 | 19 | 3022 | ||
NR_134949.1 | 20 | 3124 | ||||
NR_134950.1 | 19 | 3022 | ||||
GenBank mRNA | BC140782.1 | - | 19 | 2977 | 9021650 | |
BC144540.1 | - | 20 | 3085 | |||
BC144541.1 | - | 19 | 2977 | |||
D63395.1 | - | 2 | 159 | |||
U95299.1 | - | 19 | 2973 |