Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004589.3 | NP_004580.1 | 6 | 1667 | ||
XM_005256751.3 | XP_005256808.1 | 6 | 1634 | |||
GenBank mRNA | AF026852.1 | - | 6 | 1619 | ||
AF131816.1 | - | 5 | 1274 | |||
AK074588.1 | - | 6 | 1573 | |||
AK098826.1 | - | 6 | 1627 |