Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRAF6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004620.3 | NP_004611.1 | 3 | 708 | ||
NM_145803.2 | NP_665802.1 | 4 | 801 | |||
XM_017018220.1 | XP_016873709.1 | 3 | 708 | |||
GenBank mRNA | AK292978.1 | - | 3 | 667 | ||
BC031052.1 | - | 4 | 746 | 5272008 | ||
U78798.1 | - | 3 | 641 |