Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC33A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004733.3 | NP_004724.1 | 6 | 2451 | ||
XM_011513311.2 | XP_011511613.1 | 4 | 2118 | |||
XM_017007462.1 | XP_016862951.1 | 4 | 2115 | |||
XM_017007463.1 | XP_016862952.1 | 6 | 2534 | |||
XM_017007464.1 | XP_016862953.1 | 5 | 2220 | |||
GenBank mRNA | D88152.1 | - | 6 | 2376 |