Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NRXN1 SIRNA Assays ›
Gene Symbol:
NRXN1
Gene Name:
neurexin 1
Gene Aliases:
Hs.22998, PTHSL2, SCZD17
Chromosome Location:
Chr.2: 49918503 - 51032536 on Build GRCh38
UniGene ID:
Hs.637685
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
107542
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: NRXN1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001135659.1
NP_001129131.1
24
7468
NM_001320156.1
NP_001307085.1
3
1979
NM_001320157.1
NP_001307086.1
3
1970
NM_004801.4
NP_004792.1
22
7258
NM_138735.2
NP_620072.1
6
3483
XM_005264642.3
XP_005264699.1
23
7360
XM_005264643.3
XP_005264700.1
22
7315
XM_006712137.3
XP_006712200.1
22
7270
XM_011533167.2
XP_011531469.1
23
7369
XM_011533171.2
XP_011531473.1
23
7348
XM_011533172.2
XP_011531474.1
22
7342
XM_011533174.2
XP_011531476.1
22
7324
XM_011533175.2
XP_011531477.1
21
7312
XM_011533177.2
XP_011531479.1
22
7279
XM_011533178.2
XP_011531480.1
20
7279
XM_011533183.1
XP_011531485.1
17
5051
XM_017005303.1
XP_016860792.1
24
7396
XM_017005304.1
XP_016860793.1
24
7393
XM_017005305.1
XP_016860794.1
24
7387
XM_017005306.1
XP_016860795.1
23
7384
XM_017005307.1
XP_016860796.1
23
7378
XM_017005308.1
XP_016860797.1
24
7375
XM_017005309.1
XP_016860798.1
23
7369
XM_017005310.1
XP_016860799.1
22
7366
XM_017005311.1
XP_016860800.1
23
7351
XM_017005312.1
XP_016860801.1
23
7345
XM_017005313.1
XP_016860802.1
23
7339
XM_017005314.1
XP_016860803.1
21
7336
XM_017005315.1
XP_016860804.1
22
7333
XM_017005316.1
XP_016860805.1
22
7333
XM_017005317.1
XP_016860806.1
22
7327
XM_017005318.1
XP_016860807.1
22
7324
XM_017005319.1
XP_016860808.1
22
7321
XM_017005320.1
XP_016860809.1
22
7312
XM_017005321.1
XP_016860810.1
23
7306
XM_017005322.1
XP_016860811.1
23
7297
XM_017005323.1
XP_016860812.1
19
7282
XM_017005324.1
XP_016860813.1
21
7252
XM_017005325.1
XP_016860814.1
21
7243
XM_017005326.1
XP_016860815.1
21
7240
XM_017005327.1
XP_016860816.1
21
7225
XM_017005328.1
XP_016860817.1
20
7207
XM_017005330.1
XP_016860819.1
18
7183
XM_017005331.1
XP_016860820.1
18
7168
XM_017005332.1
XP_016860821.1
17
7147
XM_017005333.1
XP_016860822.1
17
7138
XM_017005334.1
XP_016860823.1
20
5217
XM_017005335.1
XP_016860824.1
7
8563
XM_017005336.1
XP_016860825.1
7
8555
XM_017005337.1
XP_016860826.1
6
8464
GenBank mRNA
AB011150.2
-
21
6023
AB035356.1
-
21
5959
BC150247.1
-
21
6044
9007472
BX647616.1
-
3
1925
DA153976.1
-
1
375
DA168874.1
-
1
39
EF539882.1
-
24
7331
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
neuron cell-cell adhesion
chemical synaptic transmission
neurotransmitter secretion
axon guidance
synapse assembly
learning
adult behavior
social behavior
vocal learning
neuromuscular process controlling balance
positive regulation of synapse assembly
positive regulation of synaptic transmission, glutamatergic
prepulse inhibition
regulation of insulin secretion involved in cellular response to glucose stimulus
vocalization behavior
positive regulation of synapse maturation
postsynaptic membrane assembly
gephyrin clustering involved in postsynaptic density assembly
neuroligin clustering involved in postsynaptic membrane assembly
postsynaptic density protein 95 clustering
positive regulation of excitatory postsynaptic potential
regulation of grooming behavior
receptor activity
calcium channel regulator activity
calcium ion binding
protein binding
acetylcholine receptor binding
cell adhesion molecule binding
neuroligin family protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-f2nf4:80/10.239.165.207:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0