Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CTNNA1 SIRNA Assays ›
Gene Symbol:
CTNNA1
Gene Name:
catenin alpha 1
Gene Aliases:
CAP102, MDPT2
Chromosome Location:
Chr.5: 138753386 - 138935034 on Build GRCh38
UniGene ID:
Hs.445981
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
10758
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CTNNA1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001290307.2
NP_001277236.1
16
2335
NM_001290309.2
NP_001277238.1
15
2139
NM_001290310.2
NP_001277239.1
16
2172
NM_001290312.1
NP_001277241.1
10
1655
NM_001323982.1
NP_001310911.1
17
2805
NM_001323983.1
NP_001310912.1
16
2487
NM_001323984.1
NP_001310913.1
17
2353
NM_001323985.1
NP_001310914.1
16
2335
NM_001323986.1
NP_001310915.1
15
2242
NM_001323987.1
NP_001310916.1
10
1344
NM_001323988.1
NP_001310917.1
13
1592
NM_001323989.1
NP_001310918.1
11
1440
NM_001323990.1
NP_001310919.1
12
1804
NM_001323991.1
NP_001310920.1
11
1751
NM_001323992.1
NP_001310921.1
13
1584
NM_001323993.1
NP_001310922.1
13
1898
NM_001323994.1
NP_001310923.1
10
1353
NM_001323995.1
NP_001310924.1
13
1568
NM_001323996.1
NP_001310925.1
13
1903
NM_001323997.1
NP_001310926.1
13
1587
NM_001323998.1
NP_001310927.1
13
1879
NM_001323999.1
NP_001310928.1
13
1876
NM_001324000.1
NP_001310929.1
11
1748
NM_001324001.1
NP_001310930.1
10
1655
NM_001324002.1
NP_001310931.1
10
1655
NM_001324003.1
NP_001310932.1
12
1804
NM_001324004.1
NP_001310933.1
10
1344
NM_001324005.1
NP_001310934.1
13
1879
NM_001324006.1
NP_001310935.1
14
1626
NM_001324007.1
NP_001310936.1
13
1862
NM_001324008.1
NP_001310937.1
11
1402
NM_001324009.1
NP_001310938.1
11
1713
NM_001324010.1
NP_001310939.1
7
1017
NM_001324011.1
NP_001310940.1
12
1475
NM_001324012.1
NP_001310941.1
8
1110
NM_001324013.1
NP_001310942.1
11
1684
NM_001903.4
NP_001894.2
16
2335
GenBank mRNA
AK094151.1
-
13
1544
AK296708.1
-
16
2143
AK300439.1
-
15
2106
AK304267.1
-
16
2230
BC000385.2
-
16
2273
2821111
BC031262.1
-
10
1234
5274351
D13866.1
-
16
2274
D14705.1
-
16
2299
HQ589335.1
-
10
1489
L23805.1
-
15
2215
U03100.1
-
15
2215
Back To Top
More Information
Panther Classification:
Molecular Function -
non-motor actin binding protein
Gene Ontology Categories:
Function(s)
Process(es)
ovarian follicle development
actin filament organization
cell adhesion
establishment or maintenance of cell polarity
negative regulation of neuroblast proliferation
aging
male gonad development
gap junction assembly
axon regeneration
adherens junction organization
cellular protein localization
odontogenesis of dentin-containing tooth
apical junction assembly
response to estrogen
positive regulation of smoothened signaling pathway
positive regulation of muscle cell differentiation
protein heterooligomerization
cellular response to indole-3-methanol
epithelial cell-cell adhesion
negative regulation of cell motility
negative regulation of integrin-mediated signaling pathway
negative regulation of extrinsic apoptotic signaling pathway in absence of ligand
positive regulation of extrinsic apoptotic signaling pathway in absence of ligand
structural molecule activity
protein binding
beta-catenin binding
vinculin binding
poly(A) RNA binding
gamma-catenin binding
cadherin binding
actin filament binding
cadherin binding involved in cell-cell adhesion
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-r4vqj:80/10.239.163.135:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0