Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: SCD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005063.4 | NP_005054.3 | 6 | 1594 | ||
GenBank mRNA | AB032261.1 | - | 6 | 1484 | ||
AB208982.1 | - | 6 | 1351 | |||
BC062303.1 | - | 6 | 1480 | 6586153 | ||
CD245516.1 | - | 2 | 245 | |||
Y13647.1 | - | 6 | 1312 |