Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SLC23A2 SIRNA Assays ›
Gene Symbol:
SLC23A2
Gene Name:
solute carrier family 23 member 2
Gene Aliases:
NBTL1, SLC23A1, SVCT2, YSPL2, hSVCT2
Chromosome Location:
Chr.20: 4852356 - 5010293 on Build GRCh38
UniGene ID:
Hs.516866
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
107630
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SLC23A2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_005116.5
NP_005107.4
15, 16
2007
NM_203327.1
NP_976072.1
15, 16
2025
XM_011529414.2
XP_011527716.1
15, 16
2012
XM_011529415.2
XP_011527717.1
14, 15
1833
XM_011529416.2
XP_011527718.1
14, 15
2142
XM_011529417.2
XP_011527719.1
14, 15
1889
XM_017028171.1
XP_016883660.1
15, 16
2046
XM_017028172.1
XP_016883661.1
15, 16
2050
XM_017028173.1
XP_016883662.1
15, 16
2030
XM_017028174.1
XP_016883663.1
15, 16
1925
XM_017028175.1
XP_016883664.1
11, 12
1361
XM_017028176.1
XP_016883665.1
12, 13
1679
XM_017028177.1
XP_016883666.1
12, 13
1666
XM_017028178.1
XP_016883667.1
14, 15
1932
XM_017028179.1
XP_016883668.1
14, 15
1919
XM_017028180.1
XP_016883669.1
10, 11
1243
GenBank mRNA
AF058319.1
-
13, 14
1632
AF092511.1
-
14, 15
1928
AF164142.1
-
15, 16
2025
AJ269478.1
-
13, 14
1632
AJ292318.1
-
14, 15
1863
AK296304.1
-
12, 13
1673
AY380556.1
-
13, 14
1632
D87075.2
-
15, 16
2007
EF032501.1
-
13, 14
1632
Back To Top
More Information
Panther Classification:
Molecular Function -
transporter
Gene Ontology Categories:
Function(s)
Process(es)
nucleobase-containing compound metabolic process
response to oxidative stress
nucleobase transport
L-ascorbic acid transport
molecular hydrogen transport
L-ascorbic acid metabolic process
vitamin transmembrane transport
sodium ion transmembrane transport
transepithelial L-ascorbic acid transport
L-ascorbate:sodium symporter activity
sodium-dependent multivitamin transmembrane transporter activity
nucleobase transmembrane transporter activity
L-ascorbic acid transporter activity
sodium-dependent L-ascorbate transmembrane transporter activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-xbcvc:80/10.239.164.135:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline