Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UNC119 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005148.3 | NP_005139.1 | 5 | 1059 | ||
NM_054035.2 | NP_473376.1 | 4 | 1326 | |||
XM_017025289.1 | XP_016880778.1 | 6 | 1084 | |||
GenBank mRNA | AB593014.1 | - | 4 | 1329 | ||
AF028788.1 | - | 5 | 991 | |||
AF028789.1 | - | 4 | 1259 | |||
AK124531.1 | - | 4 | 3865 | |||
BC027176.1 | - | 5 | 1033 | 4341426 | ||
BX641031.1 | - | 4 | 3788 | |||
BX647747.1 | - | 2 | 891 | |||
BX648530.1 | - | 3 | 1729 | |||
U40998.1 | - | 5 | 1042 |